3'-(Methylthio)biphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4850-5G CA-4850-1G
COBBCA-4850-5GEA 0 EUR
COBBCA-4850-5G COBBCA-4850-1G
3'-(Methylthio)biphenyl-3-carbonsäure
3'-(Methylthio)biphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₂S
MDL: MFCD06858745
CAS-Nummer: 885965-43-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR