4'-(Methylthio)biphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4848-5G
CA-4848-1G
COBBCA-4848-5GEA
0
EUR
COBBCA-4848-5G
COBBCA-4848-1G
4'-(Methylthio)biphenyl-3-carbonsäure
4'-(Methylthio)biphenyl-3-carbonsäure
Formel:
C₁₄H₁₂O₂S |
MDL:
MFCD04117399 CAS-Nummer: 728918-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...