3,5-Dichlor-4-thiocyanatoanilin

Lieferant: COMBI-BLOCKS

AN-1432-5G AN-1432-1G
COBBAN-1432-5GEA 303 EUR
COBBAN-1432-5G COBBAN-1432-1G
3,5-Dichlor-4-thiocyanatoanilin
3,5-Dichlor-4-thiocyanatoanilin
Formel: C₇H₄Cl₂N₂S
MDL: MFCD11855832
CAS-Nummer: 7494-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR