1,1-Cyclohexandiessigsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,1-Cyclohexanediaceticacid
B20816.09
B20816.18
B20816.09EA
48.2
EUR
B20816.09
B20816.18
1,1-Cyclohexandiessigsäure ≥98%
1,1-Cyclohexandiessigsäure
Purity: 98%
CAS No.: 4355-11-7
Molecular Formula: C10H16O4
Formula Weight: 200.23
Melting Point: 178-182°
MDL No.: MFCD00001519
Formel:
C₁₀H₁₆O₄ Molecular Weight: 200,23 g/mol Schmelzpunkt: 178…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001519 CAS-Nummer: 4355-11-7 EINECS: 224-427-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...