5-Brom-4-methyl-2-(4-morpholino)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 4-(5-Bromo-4-methyl-2-pyridinyl)morpholine

PY-8030-1G PY-8030-5G PY-8030-250M
COBBPY-8030-1GEA 102 EUR
COBBPY-8030-1G COBBPY-8030-5G COBBPY-8030-250M
5-Brom-4-methyl-2-(4-morpholino)pyridin
5-Brom-4-methyl-2-(4-morpholino)pyridin
Formel: C₁₀H₁₃BrN₂O
MDL: MFCD12546513
CAS-Nummer: 1187385-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR