2-(Dimethylcarbamoyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N,N-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

PN-2950-5G PN-2950-1G
COBBPN-2950-5GEA 0 EUR
COBBPN-2950-5G COBBPN-2950-1G
2-(Dimethylcarbamoyl)phenylboronsäurepinakolester
2-(Dimethylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₂BNO₃
MDL: MFCD07368236
CAS-Nummer: 956229-73-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR