3-(Diethylcarbamoyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3-(N,N-Diethylaminocarbonyl)phenylboronic acid, pinacol ester, [3-(Diethylamine-1-carbonyl)phenyl]boronic acid pinacol ester
PN-3051-5G
PN-3051-1G
COBBPN-3051-5GEA
303
EUR
COBBPN-3051-5G
COBBPN-3051-1G
3-(Diethylcarbamoyl)phenylboronsäurepinakolester
3-(Diethylcarbamoyl)phenylboronsäurepinakolester
Formel:
C₁₇H₂₆BNO₃ |
MDL:
MFCD08458202 CAS-Nummer: 325142-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...