4-(Dimethylcarbamoyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(N,N-Dimethylaminocarbonyl)phenylboronic acid, pinacol ester, 4-(Dimethylcarbamoyl)phenylboronic Acid Pinacol Ester

PN-3000-5G PN-3000-1G
COBBPN-3000-5GEA 385 EUR
COBBPN-3000-5G COBBPN-3000-1G
4-(Dimethylcarbamoyl)phenylboronsäurepinakolester
4-(Dimethylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₂BNO₃
Molecular Weight: 275.1511 g/mol
MDL: MFCD05863908
CAS-Nummer: 400727-57-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR