4-(Methylcarbamoyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

PN-3006-5G PN-3006-1G
COBBPN-3006-5GEA 370 EUR
COBBPN-3006-5G COBBPN-3006-1G
4-(Methylcarbamoyl)phenylboronsäurepinakolester
4-(Methylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₄H₂₀BNO₃
Molecular Weight: 261,1245 g/mol
MDL: MFCD06795690
CAS-Nummer: 214360-57-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR