3-Brom-2-(2'-fluorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8060-1G BB-8060-250M
COBBBB-8060-1GEA 0 EUR
COBBBB-8060-1G COBBBB-8060-250M
3-Brom-2-(2'-fluorbenzyloxy)phenylboronsäure
3-Brom-2-(2'-fluorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₁BBrFO₃
MDL: MFCD06798106
CAS-Nummer: 870778-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR