3-Brom-2-(4'-fluorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8114-1G BB-8114-250M
COBBBB-8114-1GEA 330 EUR
COBBBB-8114-1G COBBBB-8114-250M
3-Brom-2-(4'-fluorbenzyloxy)phenylboronsäure
3-Brom-2-(4'-fluorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₁BBrFO₃
MDL: MFCD06411372
CAS-Nummer: 849052-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR