4-(4-Hydroxyphenyl)benzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4'-Hydroxy-4-biphenylcarbonitrile, 4'-Hydroxybiphenyl-4-carbonitrile, 4'-Hydroxy-[1,1'-biphenyl]-4-carbonitrile

374784-1G 374784-5G
SIAL374784-1GEA 92 EUR
SIAL374784-1G SIAL374784-5G
4-(4-Hydroxyphenyl)benzonitril, Sigma-Aldrich®
4-(4-Hydroxyphenyl)benzonitril
Formel: C₁₃H₉NO
Molecular Weight: 195,22 g/mol
MDL: MFCD00059625
CAS-Nummer: 19812-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR