2,6-Difluor-3-(2'-fluorbenzyloxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-8121-1G
BB-8121-250M
COBBBB-8121-1GEA
341
EUR
COBBBB-8121-1G
COBBBB-8121-250M
2,6-Difluor-3-(2'-fluorbenzyloxy)phenylboronsäure
2,6-Difluor-3-(2'-fluorbenzyloxy)phenylboronsäure
Formel:
C₁₃H₁₀BClF₂O₃ |
MDL:
MFCD07784354 CAS-Nummer: 836615-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...