5-Brom-N-butylpicolinamid

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-N-butylpyridine-2-carboxamide

PY-7214-1G PY-7214-5G
COBBPY-7214-1GEA 149 EUR
COBBPY-7214-1G COBBPY-7214-5G
5-Brom-N-butylpicolinamid
5-Brom-N-butylpicolinamid
Formel: C₁₀H₁₃BrN₂O
MDL: MFCD09801065
CAS-Nummer: 951885-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR