5-Brom-N-ethylpicolinamid
Lieferant: COMBI-BLOCKS
Synonyme:
5-Bromo-N-ethylpyridine-2-carboxamide
PY-7178-1G
PY-7178-5G
COBBPY-7178-1GEA
149
EUR
COBBPY-7178-1G
COBBPY-7178-5G
5-Brom-N-ethylpicolinamid
5-Brom-N-ethylpicolinamid
Formel:
C₈H₉BrN₂O |
MDL:
MFCD07357422 CAS-Nummer: 845305-88-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...