5-Brom-N-methylpicolinamid

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-N-methylpyridine-2-carboxamide

PY-7177-1G PY-7177-5G
COBBPY-7177-1GEA 201 EUR
COBBPY-7177-1G COBBPY-7177-5G
5-Brom-N-methylpicolinamid
5-Brom-N-methylpicolinamid
Formel: C₇H₇BrN₂O
Molecular Weight: 215,05 g/mol
MDL: MFCD07357421
CAS-Nummer: 845305-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR