5-Brom-N-propylpicolinamid

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-N-propylpyridine-2-carboxamide

PY-7209-1G PY-7209-5G
COBBPY-7209-1GEA 204 EUR
COBBPY-7209-1G COBBPY-7209-5G
5-Brom-N-propylpicolinamid
5-Brom-N-propylpicolinamid
Formel: C₉H₁₁BrN₂O
MDL: MFCD09801061
CAS-Nummer: 845305-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR