5-Brom-N-cyclopropylpicolinamid

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-N-cyclopyridine-2-carboxamide, 5-Bromo-N-cyclopropylpyridine-2-carboxamide

PY-7210-1G PY-7210-5G
COBBPY-7210-1GEA 204 EUR
COBBPY-7210-1G COBBPY-7210-5G
5-Brom-N-cyclopropylpicolinamid
5-Brom-N-cyclopropylpicolinamid
Formel: C₉H₉BrN₂O
MDL: MFCD09801062
CAS-Nummer: 638219-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR