4-Chlor-N-methylpicolinamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-N-methylpyridine-2-carboxamide

PY-7169-1G PY-7169-5G
COBBPY-7169-1GEA 0 EUR
COBBPY-7169-1G COBBPY-7169-5G
4-Chlor-N-methylpicolinamid
4-Chlor-N-methylpicolinamid
Formel: C₇H₇ClN₂O
Molecular Weight: 170,60 g/mol
MDL: MFCD02185921
CAS-Nummer: 220000-87-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR