4-Chlorpicolinsäurehydrazid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-2-(hydrazinecarbonyl)pyridine

CA-4153-5G CA-4153-1G
COBBCA-4153-5GEA 912 EUR
COBBCA-4153-5G COBBCA-4153-1G
4-Chlorpicolinsäurehydrazid
4-Chlorpicolinsäurehydrazid
Formel: C₆H₆ClN₃O
Molecular Weight: 171,5843 g/mol
MDL: MFCD01704394
CAS-Nummer: 73771-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR