4-Methoxy-2-pyridincarbonitril

Lieferant: COMBI-BLOCKS

Synonyme: 4-methoxypicolinonitrile

CA-4161-5G CA-4161-1G
COBBCA-4161-5GEA 384 EUR
COBBCA-4161-5G COBBCA-4161-1G
4-Methoxy-2-pyridincarbonitril
4-Methoxy-2-pyridincarbonitril
Formel: C₇H₆N₂O
Molecular Weight: 134.1400 g/mol
MDL: MFCD08062857
CAS-Nummer: 36057-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR