N-Fmoc-D-valin
Lieferant: BACHEM BIOCHEMICA
Synonyme:
Fmoc-D-Val-OH, D-Valine, N-FMOC protected
4007570.0005
4007570.0025
BACE4007570.0005EA
0
EUR
BACE4007570.0005
BACE4007570.0025
N-Fmoc-D-valin
N-Fmoc-D-valin
Formel:
C₂₀H₂₁NO₄ Molecular Weight: 339,39 g/mol |
MDL:
MFCD00062953 CAS-Nummer: 84624-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...