N-Fmoc-L-valin ≥98%

Lieferant: Thermo Scientific

B21030.06 B21030.14
B21030.06EA 23.2 EUR
B21030.06 B21030.14
N-Fmoc-L-valin ≥98%
N-Fmoc-L-valin

Purity: 98% CAS No.: 68858-20-8 Molecular Formula: C20H21NO4 Formula Weight: 339.39 Storage Temperature: 2°C to 8°C Melting Point: 142-147° Beilstein Reference: 2177443 Optical Rotation: -17° (c=1 in DMF) MDL No.: MFCD00037124

Formel: C₂₀H₂₁NO₄
Molecular Weight: 339,4 g/mol
Schmelzpunkt: 142…147 °C
MDL: MFCD00037124
CAS-Nummer: 68858-20-8
EINECS: 272-515-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR