N-Fmoc-L-valin

Lieferant: BACHEM BIOCHEMICA

B-1455.0025 B-1455.0100 B-1455.0500
BACEB-1455.0025EA 0 EUR
BACEB-1455.0025 BACEB-1455.0100 BACEB-1455.0500
N-Fmoc-L-valin
N-Fmoc-L-valin
Formel: C₂₀H₂₁NO₄
Molecular Weight: 339,4 g/mol
MDL: MFCD00037124
CAS-Nummer: 68858-20-8
EINECS: 272-515-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR