N-Fmoc-β-alanin ≥99%

Lieferant: Thermo Scientific

Synonyme: Fmoc-β-Ala-OH P, Fmoc-β-Ala-p-alkoxybenzyl alcohol, N-Fmoc-beta-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-beta-alanine

H59887.06 H59887.14
H59887.06EA 62.8 EUR
H59887.06 H59887.14
N-Fmoc-β-alanin ≥99%
N-Fmoc-β-alanin
Formel: C₁₈H₁₇NO₄
Molecular Weight: 311,3 g/mol
Schmelzpunkt: 142…147 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00063328
CAS-Nummer: 35737-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR