N-Fmoc-β-alanin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Fmoc-β-Ala-OH P, Fmoc-β-Ala-p-alkoxybenzyl alcohol, N-Fmoc-beta-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-beta-alanine

47587-25G-F 47587-5G-F
SIAL47587-25G-FEA 154 EUR
SIAL47587-25G-F SIAL47587-5G-F
N-Fmoc-β-alanin, Sigma-Aldrich®
N-Fmoc-β-alanin
Formel: C₁₈H₁₇NO₄
Molecular Weight: 311,3 g/mol
MDL: MFCD00063328
CAS-Nummer: 35737-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR