(R)-3-(Fmoc-amino)-3-phenylpropionsäure 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylpropanoic acid
H52074.MD
H52074.03
H52074.MDEA
61.2
EUR
H52074.MD
H52074.03
(R)-3-(Fmoc-amino)-3-phenylpropionsäure 95%
(R)-3-(Fmoc-amino)-3-phenylpropionsäure
Purity: 95%
CAS No.: 220498-02-2
Molecular Formula: C24H21NO4
Formula Weight: 387.43
Beilstein Reference: 7982022
MDL No.: MFCD01311783
Formel:
C₂₄H₂₁NO₄ Molecular Weight: 387.43 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01311783 CAS-Nummer: 220498-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...