1-(2-(4-Nitrophenoxy)ethyl)pyrrolidin

Lieferant: COMBI-BLOCKS

AN-1191-5G AN-1191-25G AN-1191-1G
COBBAN-1191-5GEA 557 EUR
COBBAN-1191-5G COBBAN-1191-25G COBBAN-1191-1G
1-(2-(4-Nitrophenoxy)ethyl)pyrrolidin
1-(2-(4-Nitrophenoxy)ethyl)pyrrolidin
Formel: C₁₂H₁₆N₂O₃
Molecular Weight: 236.2700 g/mol
MDL: MFCD01308390
CAS-Nummer: 265654-77-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR