((4-Brom-3-methylphenyl)carbonyl)morpholin

Lieferant: COMBI-BLOCKS

CA-4566-1G CA-4566-5G
COBBCA-4566-1GEA 149 EUR
COBBCA-4566-1G COBBCA-4566-5G
((4-Brom-3-methylphenyl)carbonyl)morpholin
((4-Brom-3-methylphenyl)carbonyl)morpholin
Formel: C₁₂H₁₄BrNO₂
MDL: MFCD09972130
CAS-Nummer: 149105-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR