N-(2,6-Difluor-3-nitrobenzoyl)morpholin

Lieferant: Thermo Scientific

SEW06120.250 SEW06120.1
MAYBSEW06120.250EA 0 EUR
MAYBSEW06120.250 MAYBSEW06120.1
N-(2,6-Difluor-3-nitrobenzoyl)morpholin
N-(2,6-Difluor-3-nitrobenzoyl)morpholin
Formel: C₁₁H₁₀F₂N₂O₄
MDL: MFCD01567176
CAS-Nummer: 260554-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR