6-Methyl-4-chinazolon

Lieferant: COMBI-BLOCKS

Synonyme: 6-Methylquinazolin-4-ol, 6-Methylquinazolin-4(3H)-one

HC-6053-5G HC-6053-25G HC-6053-1G
COBBHC-6053-5GEA 312 EUR
COBBHC-6053-5G COBBHC-6053-25G COBBHC-6053-1G
6-Methyl-4-chinazolon
6-Methyl-4-chinazolon
Formel: C₉H₈N₂O
Molecular Weight: 160,1726 g/mol
MDL: MFCD00673185
CAS-Nummer: 19181-53-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR