2-Piperidino-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Piperidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(Piperidin-1-yl)pyridine-5-boronic acid pinacol ester

PN-8696-1G PN-8696-5G
COBBPN-8696-1GEA 114 EUR
COBBPN-8696-1G COBBPN-8696-5G
2-Piperidino-5-pyridinboronsäurepinakolester
2-Piperidino-5-pyridinboronsäurepinakolester
Formel: C₁₆H₂₅BN₂O₂
Molecular Weight: 288,1929 g/mol
MDL: MFCD06798274
CAS-Nummer: 852228-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR