6-Piperidino-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
BB-8696-5G
BB-8696-1G
COBBBB-8696-5GEA
0
EUR
COBBBB-8696-5G
COBBBB-8696-1G
6-Piperidino-3-pyridinboronsäure
6-Piperidino-3-pyridinboronsäure
Formel:
C₁₀H₁₅BN₂O₂ |
MDL:
MFCD06801705 CAS-Nummer: 1002129-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...