1-(6-Brompyridin-2-yl)piperidin-4-ol

Lieferant: COMBI-BLOCKS

HC-2743-5G HC-2743-25G HC-2743-1G
COBBHC-2743-5GEA 0 EUR
COBBHC-2743-5G COBBHC-2743-25G COBBHC-2743-1G
1-(6-Brompyridin-2-yl)piperidin-4-ol
1-(6-Brompyridin-2-yl)piperidin-4-ol
Formel: C₁₀H₁₃BrN₂O
MDL: MFCD09607698
CAS-Nummer: 1017781-64-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR