(-)-Cotinin ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Cotinine, (S)-1-Methyl-5-(pyridin-3-yl)pyrrolidin-2-one

L11873.03 L11873.MD
L11873.03EA 176 EUR
L11873.03 L11873.MD
(-)-Cotinin ≥98%
(-)-Cotinin

Purity: 98% CAS No.: 486-56-6 Molecular Formula: C10H12N2O Formula Weight: 176.22 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 34-39° Boiling Point: 145-147°/3mm Beilstein Reference: 83099 Merck Reference: 14,2553 Optical Rotation: -20° (c=1 in ethanol) MDL No.: MFCD00077696

Formel: C₁₀H₁₂N₂O
Molecular Weight: 176,22 g/mol
Siedepunkt: 145…147 °C (3 mmHg)
Schmelzpunkt: 34…39 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00077696
CAS-Nummer: 486-56-6
EINECS: 207-634-9
Merck Index: 14,2553

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR