(-)-Cotinin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cotinine, (S)-1-Methyl-5-(pyridin-3-yl)pyrrolidin-2-one

M-Clarity™ Qualitätsstufe = MQ200

C5923-1G C5923-250MG
SIALC5923-1GEA 429 EUR
SIALC5923-1G SIALC5923-250MG
(-)-Cotinin, Sigma-Aldrich®
(-)-Cotinin
Formel: C₁₀H₁₂N₂O
Molecular Weight: 176.22 g/mol
MDL: MFCD00077696
CAS-Nummer: 486-56-6
EINECS: 207-634-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR