Kalium(1-methyl-4-trifluoroboratomethyl)piperazin

Lieferant: COMBI-BLOCKS

Synonyme: Potassium trifluoro((4-methylpiperazin-1-yl)methyl)borate

TB-0010-5G TB-0010-1G
COBBTB-0010-5GEA 312 EUR
COBBTB-0010-5G COBBTB-0010-1G
Kalium(1-methyl-4-trifluoroboratomethyl)piperazin
Kalium(1-methyl-4-trifluoroboratomethyl)piperazin
Formel: C₆H₁₃BF₃KN₂
Molecular Weight: 220,0853 g/mol
MDL: MFCD10700160
CAS-Nummer: 1015484-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR