Ethyltetrahydropyran-4-ylacetat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-(tetrahydro-2H-pyran-4-yl)acetate

AM-1007-5G AM-1007-1G
COBBAM-1007-5GEA 273 EUR
COBBAM-1007-5G COBBAM-1007-1G
Ethyltetrahydropyran-4-ylacetat
Ethyltetrahydropyran-4-ylacetat
Formel: C₉H₁₆O₃
Molecular Weight: 172,2215 g/mol
MDL: MFCD03788560
CAS-Nummer: 103260-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR