Methyltetrahydropyran-4-ylacetat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-(tetrahydro-2H-pyran-4-yl)acetate

AM-1629-1G AM-1629-25G AM-1629-5G
COBBAM-1629-1GEA 127 EUR
COBBAM-1629-1G COBBAM-1629-25G COBBAM-1629-5G
Methyltetrahydropyran-4-ylacetat
Methyltetrahydropyran-4-ylacetat
Formel: C₈H₁₄O₃
Molecular Weight: 158.2000 g/mol
MDL: MFCD09972087
CAS-Nummer: 156002-64-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR