4-[5-Chlor-2-nitro-4-(trifluormethyl)phenyl]-2,6-dimethylmorpholin

Lieferant: Thermo Scientific

BTB02418.250 BTB02418.1
MAYBBTB02418.250EA 0 EUR
MAYBBTB02418.250 MAYBBTB02418.1
4-[5-Chlor-2-nitro-4-(trifluormethyl)phenyl]-2,6-dimethylmorpholin
4-[5-Chlor-2-nitro-4-(trifluormethyl)phenyl]-2,6-dimethylmorpholin
Formel: C₁₃H₁₄ClF₃N₂O₃
MDL: MFCD00204831
CAS-Nummer: 646456-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR