4-(2,6-Dimethylmorpholino)anilin

Lieferant: BIONET RESEARCH (CHEMPUR)

BIOT9W-0242.5EA 0 EUR
BIOT9W-0242.5 BIOT9W-0242.10 BIOT9W-0242.500 BIOT9W-0242.1
4-(2,6-Dimethylmorpholino)anilin
4-(2,6-Dimethylmorpholino)anilin
Formel: C₁₂H₁₈N₂O
MDL: MFCD00051574
CAS-Nummer: 218930-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR