4-(2,6-Dimethylmorpholino)anilin
Lieferant: Thermo Scientific
BTB04213.1
BTB04213.10
MAYBBTB04213.1EA
0
EUR
MAYBBTB04213.1
MAYBBTB04213.10
4-(2,6-Dimethylmorpholino)anilin
4-(2,6-Dimethylmorpholino)anilin
Formel:
C₁₂H₁₈N₂O |
MDL:
MFCD00051574 CAS-Nummer: 218930-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...