2-(Imidazol-1-yl)-3-cyanpyridin

Lieferant: COMBI-BLOCKS

PY-8041-1G PY-8041-5G
COBBPY-8041-1GEA 0 EUR
COBBPY-8041-1G COBBPY-8041-5G
2-(Imidazol-1-yl)-3-cyanpyridin
2-(Imidazol-1-yl)-3-cyanpyridin
Formel: C₉H₆N₄
MDL: MFCD09863416
CAS-Nummer: 94448-88-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR