2-Chlor-6-(5-(trifluormethyl)-1H-pyrazol-1-yl)pyridin

Lieferant: COMBI-BLOCKS

HC-2479-5G HC-2479-1G
COBBHC-2479-5GEA 0 EUR
COBBHC-2479-5G COBBHC-2479-1G
2-Chlor-6-(5-(trifluormethyl)-1H-pyrazol-1-yl)pyridin
2-Chlor-6-(5-(trifluormethyl)-1H-pyrazol-1-yl)pyridin
Formel: C₉H₅ClF₃N₃
MDL: MFCD12026018
CAS-Nummer: 1150164-45-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR