1-(4-Bromphenyl)-2,5-dimethylpyrrol

Lieferant: BIONET RESEARCH

Synonyme: 1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrole

8W-0354.500 8W-0354.1 8W-0354.5 8W-0354.10
BIOT8W-0354.500EA 0 EUR
BIOT8W-0354.500 BIOT8W-0354.1 BIOT8W-0354.5 BIOT8W-0354.10
1-(4-Bromphenyl)-2,5-dimethylpyrrol
1-(4-Bromophenyl)-2,5-dimethylpyrrole
Formel: C₁₂H₁₂BrN
Molecular Weight: 250.14 g/mol
MDL: MFCD00277042
CAS-Nummer: 5044-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR