4-(Furfurylcarbamoyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-(Furan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide, 4-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester

PN-3019-5G PN-3019-1G
COBBPN-3019-5GEA 343 EUR
COBBPN-3019-5G COBBPN-3019-1G
4-(Furfurylcarbamoyl)phenylboronsäurepinakolester
4-(Furfurylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₈H₂₂BNO₄
Molecular Weight: 327,18 g/mol
MDL: MFCD09266168
CAS-Nummer: 1073353-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR