4-(Furfurylcarbamoyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(Furan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide, 4-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

706183-1G 706183-5G
SIAL706183-1GEA 84.8 EUR
SIAL706183-1G SIAL706183-5G
4-(Furfurylcarbamoyl)phenylboronsäurepinakolester, Sigma-Aldrich®
4-(Furfurylcarbamoyl)phenylboronsäurepinakolester
Formel: C₁₈H₂₂BNO₄
Molecular Weight: 327,18 g/mol
MDL: MFCD09266168
CAS-Nummer: 1073353-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR