2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoesäure

Lieferant: FLUOROCHEM

017512-2.5G 017512-10G 017512-1G
FLUO017512-2.5GEA 0 EUR
FLUO017512-2.5G FLUO017512-10G FLUO017512-1G
2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoesäure
2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoic acid
Formel: C₁₂H₁₂N₂O₂
MDL: MFCD02682016
CAS-Nummer: 321309-43-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR