4-(2-Thienyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(Thiophen-2-yl)phenyl)boronic acid

BB-9044-1G BB-9044-250M
COBBBB-9044-1GEA 497 EUR
COBBBB-9044-1G COBBBB-9044-250M
4-(2-Thienyl)phenylboronsäure
4-(2-Thienyl)phenylboronsäure
Formel: C₁₀H₉BO₂S
Molecular Weight: 204.0500 g/mol
MDL: MFCD06797461
CAS-Nummer: 362612-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR