Uracil-5-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2,4-Dihydroxypyrimidine-5-boronic acid, (2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)boronic acid

BB-8330-1G BB-8330-250M
COBBBB-8330-1GEA 2150 EUR
COBBBB-8330-1G COBBBB-8330-250M
Uracil-5-boronsäure
Uracil-5-boronsäure
Formel: C₄H₅BN₂O₄
Molecular Weight: 155.90 g/mol
MDL: MFCD01318983
CAS-Nummer: 70523-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR